Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ FKBP14 Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

$293.00 - $713.00

Specifications

Common Name FKBP14
Conjugate Unlabeled
For Use With (Application) ELISA, Functional Assay, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14526901
View Documents
Novus Biologicals™
NBP1452690.1MG
0.1mg
Each for $293.00
Add to cart
 
NBP1452695
View Documents
Novus Biologicals™
NBP1452690.5MG
0.5mg
Each for $713.00
Add to cart
 
Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 20-211 of Human FKBP14 The Recombinant Human FKBP14 Protein is derived from E. coli. The Recombinant Human FKBP14 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

FKBP14
ELISA, Functional Assay, SDS-Page
RUO
Unlabeled
Store at -80°C. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit