Learn More
Novus Biologicals™ Follistatin-related Gene Protein/FLRG/Fstl3 Overexpression Lysate
Supplier: Novus Biologicals™ NBL110848
Description
The Follistatin-related Gene Protein/FLRG/Fstl3 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
Western Blot | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
The lysate was created in HEK293T cells, using plasmid ID RC2811 and based on accession number NM_586. The protein contains a C-terminal DDK tag. | |
RIPA buffer | |
Homo sapiens follistatin-like 3 (secreted glycoprotein) (FSTL3), mRNA. | |
Native |