Learn More
Novus Biologicals™ g-Linked ATP Agarose Affinity Resin
Affinity resin for purification of enzymes that bind nucleotides and related molecules
$1062.00
Specifications
Product Type g-Linked ATP Agarose Affinity Resin
View More Specs
Products 2
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
51-000-02
View Documents
Novus Biologicals™
5100002
High ligand substitution (8-12μmol/mL) 2mL
Each for $1,062.00
Add to cart
51-000-05
View Documents
Novus Biologicals™
5100005
High ligand substitution (8-12μmol/mL) 5mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Resins in which ATP is linked via the gamma-phosphate have been valuable in identifying proteins in the purine-binding proteome, which includes kinases, heat shock proteins and other ATP-binding proteins.
  • ATP Agarose resin comprises ATP attached to agarose beads via its gamma-phosphate
  • Two forms of resin are available with low and high ligand substitution
  • Long hydrophilic spacer (14-atom) minimises unwanted hydrophobic interactions and facilitates unhindered interactions with biomolecules
Specifications
Specifications
g-Linked ATP Agarose Affinity Resin