Learn More
Novus Biologicals™ g-Linked GTP Agarose Affinity Resin
Affinity resin for purification of enzymes that bind nucleotides and related molecules
$1574.00
Specifications
Product Type g-Linked GTP Agarose Affinity Resin
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-500-02K
View Documents
Novus Biologicals™
5050002
5mL
Each for $1,574.00
Add to cart
50-500-01K
View Documents
Novus Biologicals™
5050001
2mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • GTP Agarose resin comprises GTP attached to agarose beads via its gamma-phosphate
  • Resin is resistant to phosphatases found in many crude tissue extracts
  • Long hydrophilic spacer (14-atom) minimises unwanted hydrophobic interactions and facilitates unhindered interactions with biomolecules
Specifications
Specifications
g-Linked GTP Agarose Affinity Resin