Learn More
Novus Biologicals™ Glucose 6 Phosphate Dehydrogenase Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130178
Catalog No. NBP130178
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1- 558 of Human Glucose 6 Phosphate Dehydrogenase The Recombinant Human Glucose 6 Phosphate Dehydrogenase Protein is derived from E. coli. The Recombinant Human Glucose 6 Phosphate Dehydrogenase Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_000166
20mM Tris-HCl buffer (pH8.0), 1mM DTT, 10% glycerol
65.3kDa
Protein
MGSSHHHHHH SSGLVPRGSH MAALTRDPQF QKLQQWYREH RSELNLRRLF DANKDRFNHF SLTLNTNHGH ILVDYSKNLV TEDVMRMLVD LAKSRGVEAA RERMFNGEKI NYTEGRAVLH VALRNRSNTP ILVDGKDVMP EVNKVLDKMK SFCQRVRSGD WKGYTGKTIT DVINIGIGGS DLGPLMVTEA LKPYSSGGPR VWYVSNIDGT HIAKTLAQLN PESSLFIIAS KTFTTQETIT NAETAKEWFL QAAKDPSAVA KHFVALSTNT TKVKEFGIDP QNMFEFWDWV GGRYSLWSAI GLSIALHVGF DNFEQLLSGA HWMDQHFRTT PLEKNAPVLL ALLGIWYINC FGCETHAMLP YDQYLHRFAA YFQQGDMESN GKYITKSGTR VDHQTGPIVW GEPGTNGQHA FYQLIHQGTK MIPCDFLIPV QTQHPIRKGL HHKILLANFL AQTEALMRGK STEEARKELQ AAGKSPEDLE RLLPHKVFEG NRPTNSIVFT KLTPFMLGAL VAMYEHKIFV QGIIWDINSF DQWGVELGKQ LAKKIEPELD GSAQVTSHDA STNGLINFIK QQREARVQ
ELISA, SDS-Page
2539
Glucose 6 Phosphate Dehydrogenase Protein
>95%
−80°C. Avoid freeze-thaw cycles.