Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HDHD1A Protein

Highly purified. Generating reliable and reproducible results.

$320.85 - $895.85

Specifications

Common Name HDHD1A
Conjugate Unlabeled
For Use With (Application) SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15088301
View Documents
Novus Biologicals™
NBP1508830.1MG
0.1mg Each for $320.85
Add to cart
 
NBP1508835
View Documents
Novus Biologicals™
NBP1508830.5MG
0.5mg Each for $895.85
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-228 of Human HDHD1A The Recombinant Human HDHD1A Protein is derived from E. coli. The Recombinant Human HDHD1A Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

HDHD1A
SDS-Page
RUO
Unlabeled
Store at -80°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit