Learn More
Novus Biologicals™ Histone H2B, ac Lys34, ac Lys43, Peptide, Biotin
Highly purified. Generating reliable and reproducible results. Applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize
Manufacturer:  Novus Biologicals™ NBP1946881.0MG
 View more versions of this product
Catalog No. NBP19468810
Add to cart
Description
Description
0 The Histone H2B [ac Lys34, ac Lys43] Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.
Specifications
Specifications
440689
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Biot-Ahx-KKRKRSR[K-Ac]ESYSVYVY[K-Ac]VLKQ-OH
Store at -80°C. Avoid freeze-thaw cycles.
Histone H2B
1mg
Lyophilized. Reconstitute in diH20.
RUO
Biotin