Learn More
Novus Biologicals™ Histone H2B, ac Lys34, Peptide, Biotin
Highly purified. Generating reliable and reproducible results. Applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize
Manufacturer:  Novus Biologicals™ NBP1946850.5MG
 View more versions of this product
Catalog No. NBP1946855
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
440689
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Biot-Ahx-KKRKRSR[K-Ac]ESYSVYVYKVLKQ-OH
Store at -80°C. Avoid freeze-thaw cycles.
Histone H2B
0.5mg
Lyophilized. Reconstitute in diH20.
RUO
Biotin