Learn More
Novus Biologicals™ Histone H2B Peptide (non-modified a.a. 27-47), Biotin
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP1946841.0MG
 View more versions of this product
Catalog No. NBP19468410
Add to cart
Description
Description
non-modified a.a. 27-47 The Histone H2B Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.
Specifications
Specifications
440689
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Biot-Ahx-KKRKRSRKESYSVYVYKVLKQ-OH
Store at -80°C. Avoid freeze-thaw cycles.
Histone H2B
1mg
Lyophilized. Reconstitute in diH20.
RUO
Biotin