Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Novus Biologicals™ Histone H3, Dimethyl Lys36, Peptide
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP1946150.5MG
 View more versions of this product
Catalog No. NBP1946155
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
126961
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Ac-SAPATGGV[K-Me2]KPHRYRPG-amide
Store at -80°C. Avoid freeze-thaw cycles.
Histone H3
0.5mg
Lyophilized. Reconstitute in diH20.
RUO
Unlabeled