Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Histone H3, Monomethyl Arg17, Peptide

Highly purified. Generating reliable and reproducible results. Applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize

$314.00

Specifications

Gene ID (Entrez) 126961
For Use With (Application) Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP19457410
View Documents
Novus Biologicals™
NBP1945741.0MG
1mg
Each for $314.00
Add to cart
 
Description

Description

0 The Histone H3 [Monomethyl Arg17] Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.
Specifications

Specifications

126961
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Store at -80°C. Avoid freeze-thaw cycles.
RUO
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit