Learn More
Novus Biologicals™ Histone H3, Monomethyl Lys36, Peptide, Biotin
Manufacturer:  Novus Biologicals™ NBP1946091.0MG
 View more versions of this product
Catalog No. NBP19460910
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
126961
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
ATKAARKSAPATGGV[K-Me1]KPHRYRPGG[K-Ahx-Biot]-OH
Store at -80°C. Avoid freeze-thaw cycles.
Histone H3
1mg
Lyophilized. Reconstitute in diH20.
RUO
Biotin