Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Novus Biologicals™ Histone H3, Monomethyl Lys36, Peptide, Biotin
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP1946061.0MG
 View more versions of this product
Catalog No. NBP19460610
Description
Description
The Histone H3 [Monomethyl Lys36] Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.
Specifications
Specifications
126961
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Ac-ATKAARKSAPATGGV[KMe1]KPHRYRPGG[K-Ahx-Biot]-amide
Store at -80°C. Avoid freeze-thaw cycles.
Histone H3
1mg
Lyophilized. Reconstitute in diH20.
RUO
Biotin