Learn More
Novus Biologicals™ Histone H3 Peptide (non-modified a.a. 1-15), Biotin
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP1945311.0MG
 View more versions of this product
Catalog No. NBP19453110
Add to cart
Description
Description
non-modified a.a. 1-15 The Histone H3 Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.
Specifications
Specifications
126961
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
ARTKQTARKSTGGKA[K-Ahx-Biot]-amide
Store at -80°C. Avoid freeze-thaw cycles.
Histone H3
1mg
Lyophilized. Reconstitute in diH20.
RUO
Biotin