Learn More
Novus Biologicals™ Histone H3 Peptide (non-modified a.a. 1-21)
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP1945461.0MG
 View more versions of this product
Catalog No. NBP19454610
Add to cart
Description
Description
non-modified a.a. 1-21 The Histone H3 Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.
Specifications
Specifications
126961
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
ARTKQTARKSTGGKAPRKQLA-amide
Store at -80°C. Avoid freeze-thaw cycles.
Histone H3
1mg
Lyophilized. Reconstitute in diH20.
RUO
Unlabeled