Learn More
Novus Biologicals™ Histone H4, Dimethyl Lys20, Peptide
Manufacturer:  Novus Biologicals™ NBP1946650.5MG
 View more versions of this product
Catalog No. NBP1946655
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
121504
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Ac-KGGAKRHR[K-Me2]VLRDNIQG-Ahx-C-amide
Store at -80°C. Avoid freeze-thaw cycles.
Histone H4
0.5mg
Lyophilized. Reconstitute in diH20.
RUO
Unlabeled