Learn More
Novus Biologicals™ Histone H4, Dimethyl Lys5, Peptide, Biotin
Highly purified. Generating reliable and reproducible results. Applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize
Manufacturer:  Novus Biologicals™ NBP1946521.0MG
 View more versions of this product
Catalog No. NBP19465210
Add to cart
Description
Description
0 The Histone H4 [Dimethyl Lys5] Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.
Specifications
Specifications
121504
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
SGRG[K-Me2]GGKGLGKGGAKRHRKVLRDNG[K-Ahx-Biot]-amide
Store at -80°C. Avoid freeze-thaw cycles.
Histone H4
1mg
Lyophilized. Reconstitute in diH20.
RUO
Biotin