Learn More
Novus Biologicals™ Histone H4 Peptide (non-modified a.a. 1-25), Biotin
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP1946441.0MG
 View more versions of this product
Catalog No. NBP19464410
Add to cart
Description
Description
non-modified a.a. 1-25 The Histone H4 Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.
Specifications
Specifications
121504
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
SGRGKGGKGLGKGGAKRHRKVLRDNG[K-Biot-Ahx]-amide
Store at -80°C. Avoid freeze-thaw cycles.
Histone H4
1mg
Lyophilized. Reconstitute in diH20.
RUO
Biotin