Learn More
Novus Biologicals™ Histone H4, Sym-Dimethyl Arg3, Peptide
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP1946391.0MG
 View more versions of this product
Catalog No. NBP19463910
Add to cart
Description
Description
The Histone H4 [Sym-dimethyl Arg3] Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.
Specifications
Specifications
121504
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Ac-SG[R-Me2s]GKGGKGLGKGGAK-Ahx-C-amide
Store at -80°C. Avoid freeze-thaw cycles.
Histone H4
1mg
Lyophilized. Reconstitute in diH20.
RUO
Unlabeled