Learn More
Novus Biologicals™ Histone H4, trimethyl Lys44, Peptide
Highly purified. Generating reliable and reproducible results. Applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize
Manufacturer:  Novus Biologicals™ NBP1946831.0MG
 View more versions of this product
Catalog No. NBP19468310
Add to cart
Description
Description
0 The Histone H4 [Trimethyl Lys44] Peptide has been validated for the following applications: ELISA, Functional, Gel Super Shift Assays, Radioimmunoassay, Peptide ELISA, Dot Blot, Block/Neutralize.
Specifications
Specifications
121504
Blocking, Neutralization, Dot Blot, ELISA, Gel Super Shift, Peptide ELISA, Radioimmunoassay
Ac-RLARRGGV[K-Me3]RISGLIYE-Ahx-C-amide
Store at -80°C. Avoid freeze-thaw cycles.
Histone H4
1mg
Lyophilized. Reconstitute in diH20.
RUO
Unlabeled