Learn More
Novus Biologicals™ Human Aldehyde Dehydrogenase 1-A1/ALDH1A1 Recombinant Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130263
Catalog No. NBP130263
Add to cart
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 1-501 of Human Aldehyde Dehydrogenase 1-A1/ALDH1A1 The Recombinant Human Aldehyde Dehydrogenase 1-A1/ALDH1A1 Protein is derived from E. coli. The Recombinant Human Aldehyde Dehydrogenase 1-A1/ALDH1A1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
P00352
50mM Tris-HCl buffer (pH7.5), 10% glycerol
54.8kDa
Protein
0.1mg
Aldehyde dehydrogenase 1A1, 1-501aa, Human, E.coli (NP_000680).
RUO
Human
WB, ELISA
216
Aldehyde Dehydrogenase 1-A1/ALDH1A1 Protein
>90%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes