Learn More
Novus Biologicals™ Human Aldehyde Dehydrogenase 3-A1/ALDH3A1 Recombinant Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
Manufacturer:  Novus Biologicals™ NBP137091
Catalog No. NBP137091
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-453 of Human Aldehyde Dehydrogenase 3-A1/ALDH3A1 The Recombinant Human Aldehyde Dehydrogenase 3-A1/ALDH3A1 Protein is derived from E. coli. The Recombinant Human Aldehyde Dehydrogenase 3-A1/ALDH3A1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications
Specifications
AAH04370
20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.1 M NaCl
52.5kDa
Protein
0.05mg
1-453aa MGSSHHHHHH SSGLVPRGSH MSKISEAVKR ARAAFSSGRT RPLQFRIQQL EALQRLIQEQ EQELVGALAA DLHKNEWNAY YEEVVYVLEE IEYMIQKLPE WAADEPVEKT PQTQQDELYI HSEPLGVVLV IGTWNYPFNL TIQPMVGAIA AGNAVVLKPS ELSENMASLL ATIIPQYLDK DLYPVINGGV PETTELLKER FDHILYTGST GVGKIIMTAA AKHLTPVTLE LGGKSPCYVD KNCDLDVACR RIAWGKFMNS GQTCVAPDYI LCDPSIQNQI VEKLKKSLKE FYGEDAKKSR DYGRIISARH FQRVMGLIEG QKVAYGGTGD AATRYIAPTI LTDVDPQSPV MQEEIFGPVL PIVCVRSLEE AIQFINQREK PLALYMFSSN DKVIKKMIAE TSSGGVAAND VIVHITLHSL PFGGVGNSGM GSYHGKKSFE TFSHRRSCLV RPLMNDEGLK VRYPPSPAKM TQH
RUO
Human
WB
218
Aldehyde Dehydrogenase 3-A1/ALDH3A1 Protein
>95%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes