Learn More
Novus Biologicals™ Human Aldo-keto Reductase 1C1/AKR1C1 Recombinant Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
Manufacturer:  Novus Biologicals™ NBP141223
Catalog No. NBP141223
Add to cart
Description
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-323 of Human Aldo-keto Reductase 1C1/AKR1C1 The Recombinant Human Aldo-keto Reductase 1C1/AKR1C1 Protein is derived from E. coli. The Recombinant Human Aldo-keto Reductase 1C1/AKR1C1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications
Specifications
NP_001344
20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 20% glycerol
38.9kDa
Protein
0.1mg
1-323aa Human, His-tagged, MGSSHHHHHH SSGLVPRGSH MDSKYQCVKL NDGHFMPVLG FGTYAPAEVP KSKALEATKL AIEAGFRHID SAHLYNNEEQ VGLAIRSKIA DGSVKREDIF YTSKLWCNSH RPELVRPALE RSLKNLQLDY VDLYLIHFPV SVKPGEEVIP KDENGKILFD TVDLCATWEA VEKCKDAGLA KSIGVSNFNR RQLEMILNKP GLKYKPVCNQ VECHPYFNQR KLLDFCKSKD IVLVAYSALG SHREEPWVDP NSPVLLEDPV LCALAKKHKR TPALIALRYQ LQRGVVVLAK SYNEQRIRQN VQVFEFQLTS EEMKAIDGLN RNVRYLTLDI FAGPPNYPFS DEY
RUO
Human
WB, ELISA
1645
Aldo-keto Reductase 1C1/AKR1C1 Protein
>90%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes