Learn More
Novus Biologicals™ Human Annexin A3 Recombinant Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP137072
Catalog No. NBP137072
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 1-323 of Human Annexin A3 The Recombinant Human Annexin A3 Protein is derived from E. coli. The Recombinant Human Annexin A3 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_005130
20mM Tris-HCl buffer (pH7.5), 1mM DTT, 10% glycerol
36.3kDa
Protein
0.1mg
Full length Annexin A3, 1-323a, MASIWVGHRGTVRDYPDFSPSVDAEAIQKAIRGIGTDEKMLISILTERSNAQRQLIVKEYQAAYGKELKDDLKGDLSGHFEHLMVALVTPPAVFDAKQLKKSMKGAGTNEDALIEILTTRTSRQMKDISQAYYTVYKKSLGDDISSETSGDFRKALLTLADGRRDESLKVDEHLAKQDAQILYKAGENRWGTDEDKFTEILCLRSFPQLKLTFDEYRNISQKDIVDSIKGELSGHFEDLLLAIVNCVRNTPAFLAERLHRALKGIGTDEFTLNRIMVSRSEIDLLDIRTEFKKHYGYSLYSAIKSDTSGDYEITLLKICGGDD
RUO
Human
ELISA, SDS-Page
306
Annexin A3 Protein
>90%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes