Learn More
Novus Biologicals™ Human Annexin A6 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP137078
Catalog No. NBP137078
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-673 of Human Annexin A6 The Recombinant Human Annexin A6 Protein is derived from E. coli. The Recombinant Human Annexin A6 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_001146
20mM Tris-HCl buffer (pH8.0), 10% glycerol, 0.1 M NaCl
78kDa
Protein
MGSSHHHHHH SSGLVPRGSH MAKPAQGAKY RGSIHDFPGF DPNQDAEALY TAMKGFGSDK EAILDIITSR SNRQRQEVCQ SYKSLYGKDL IADLKYELTG KFERLIVGLM RPPAYCDAKE IKDAISGIGT DEKCLIEILA SRTNEQMHQL VAAYKDAYER DLEADIIGDT SGHFQKMLVV LLQGTREEDD VVSEDLVQQD VQDLYEAGEL KWGTDEAQFI YILGNRSKQH LRLVFDEYLK TTGKPIEASI RGELSGDFEK LMLAVVKCIR STPEYFAERL FKAMKGLGTR DNTLIRIMVS RSELDMLDIR EIFRTKYEKS LYSMIKNDTS GEYKKTLLKL SGGDDDAAGQ FFPEAAQVAY QMWELSAVAR VELKGTVRPA NDFNPDADAK ALRKAMKGLG TDEDTIIDII THRSNVQRQQ IRQTFKSHFG RDLMTDLKSE ISGDLARLIL GLMMPPAHYD AKQLKKAMEG AGTDEKALIE ILATRTNAEI RAINEAYKED YHKSLEDALS SDTSGHFRRI LISLATGHRE EGGENLDQAR EDAQVAAEIL EIADTPSGDK TSLETRFMTI LCTRSYPHLR RVFQEFIKMT NYDVEHTIKK EMSGDVRDAF VAIVQSVKNK PLFFADKLYK SMKGAGTDEK TLTRIMVSRS EIDLLNIRRE FIEKYDKSLH QAIEGDTSGD FLKALLALCG GED
Human
ELISA, SDS-Page
309
Annexin A6 Protein
>90%
−80°C. Avoid freeze-thaw cycles.