Learn More
Novus Biologicals™ Human Apolipoprotein A-I/ApoA1 Recombinant Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130197
Catalog No. NBP130197
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 25-267 of Human Apolipoprotein A-I/ApoA1 The Recombinant Human Apolipoprotein A-I/ApoA1 Protein is derived from E. coli. The Recombinant Human Apolipoprotein A-I/ApoA1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_000030
20mM Tris-HCl buffer (pH8.0), 10% glycerol
30.3kDa
Protein
0.1mg
Apolipoprotein A-1, 25-267 aa, Human, His-tagged, Recombinant, E.coli (NP_000030).
RUO
Human
SDS-Page
335
Apolipoprotein A-I/ApoA1 protein
>95%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes