Learn More
Novus Biologicals™ Human Apolipoprotein M/ApoM Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP141151
Catalog No. NBP141151
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 23-188 of Human Apolipoprotein M/ApoM The Recombinant Human Apolipoprotein M/ApoM Protein is derived from E. coli. The Recombinant Human Apolipoprotein M/ApoM Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_061974
20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT
20.9kDa
Protein
MGSSHHHHHH SSGLVPRGSH MCPEHSQLTT LGVDGKEFPE VHLGQWYFIA GAAPTKEELA TFDPVDNIVF NMAAGSAPMQ LHLRATIRMK DGLCVPRKWI YHLTEGSTDL RTEGRPDMKT ELFSSSCPGG IMLNETGQGY QRFLLYNRSP HPPEKCVEEF KSLTSCLDSK AFLLTPRNQE ACELSNN
Human
ELISA, SDS-Page
55937
Apolipoprotein M/ApoM Protein
>90%
−80°C. Avoid freeze-thaw cycles.