Learn More
Novus Biologicals™ Human BCL-W/BCL2L2 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP137073
Catalog No. NBP137073
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-172 of Human BCL-W/BCL2L2 The Recombinant Human BCL-W/BCL2L2 Protein is derived from E. coli. The Recombinant Human BCL-W/BCL2L2 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_004041
20mM Tris-HCl buffer (pH7.5), 100mM NaCl, 10% glycerol
15.1kDa
Protein
MGSSHHHHHH SSGLVPRGSH MATPASAPDT RALVADFVGY KLRQKGYVCG AGPGEGPAAD PLHQAMRAAG DEFETRFRRT FSDLAAQLHV TPGSAQQRFT QVSDELFQGG PNWGRLVAFF VFGAALCAES VNKEMEPLVG QVQEWMVAYL ETRLADWIHS SGGWAEFTAL YGDGALEEAR RLREGNWASV RT
Human
ELISA, SDS-Page
599
BCL-W/BCL2L2 Protein
>95%
−80°C. Avoid freeze-thaw cycles.