Learn More
Novus Biologicals™ Human Biliverdin Reductase A/BLVRA Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130216
Catalog No. NBP130216
Add to cart
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 3-296 of Human Biliverdin Reductase A/BLVRA The Recombinant Human Biliverdin Reductase A/BLVRA Protein is derived from E. coli. The Recombinant Human Biliverdin Reductase A/BLVRA Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications
Specifications
NP_000703
20mM Tris-HCl buffer (pH8.0), 10% glycerol
33.3kDa
Protein
MAEPERKFGV VVVGVGRAGS VRMRDLRNPH PSSAFLNLIG FVSRRELGSI DGVQQISLED ALSSQEVEVA YICSESSSHE DYIRQFLNAG KHVLVEYPMT LSLAAAQELW ELAEQKGKVL HEEHVELLME EFAFLKKEVV GKDLLKGSLL FTAGPLEEER FGFPAFSGIS RLTWLVSLFG ELSLVSATLE ERKEDQYMKM TVCLETEKKS PLSWIEEKGP GLKRNRYLSF HFKSGSLENV PNVGVNKNIF LKDQNIFVQK LLGQFSEKEL AAEKKRILHC LGLAEEIQKY CCSRK
Human
ELISA, SDS-Page
644
Biliverdin Reductase A/BLVRA Protein
>90%
−80°C. Avoid freeze-thaw cycles.