Learn More
Novus Biologicals™ Human Carbonic Anhydrase I/CA1 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130290
Catalog No. NBP130290
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-261 of Human Carbonic Anhydrase I/CA1 The Recombinant Human Carbonic Anhydrase I/CA1 Protein is derived from E. coli. The Recombinant Human Carbonic Anhydrase I/CA1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_001122301
20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol
31kDa
Protein
MGSSHHHHHH SSGLVPRGSH MASPDWGYDD KNGPEQWSKL YPIANGNNQS PVDIKTSETK HDTSLKPISV SYNPATAKEI INVGHSFHVN FEDNDNRSVL KGGPFSDSYR LFQFHFHWGS TNEHGSEHTV DGVKYSAELH VAHWNSAKYS SLAEAASKAD GLAVIGVLMK VGEANPKLQK VLDALQAIKT KGKRAPFTNF DPSTLLPSSL DFWTYPGSLT HPPLYESVTW IICKESISVS SEQLAQFRSL LSNVEGDNAV PMQHNNRPTQ PLKGRTVRAS F
Human
ELISA, SDS-Page
759
Carbonic Anhydrase I/CA1 protein
>95%
−80°C. Avoid freeze-thaw cycles.