Learn More
Novus Biologicals™ Human Carbonic Anhydrase III/CA3 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP141230
Catalog No. NBP141230
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 1-260 of Human Carbonic Anhydrase III/CA3 The Recombinant Human Carbonic Anhydrase III/CA3 Protein is derived from E. coli. The Recombinant Human Carbonic Anhydrase III/CA3 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
AAH04897
20mM Tris-HCl buffer (pH8.0), 10% glycerol, 1mM DTT
29.5kDa
Protein
MAKEWGYASH NGPDHWHELF PNAKGENQSP IELHTKDIRH DPSLQPWSVS YDGGSAKTIL NNGKTCRVVF DDTYDRSMLR GGPLPGPYRL RQFHLHWGSS DDHGSEHTVD GVKYAAELHL VHWNPKYNTF KEALKQRDGI AVIGIFLKIG HENGEFQIFL DALDKIKTKG KEAPFTKFDP SCLFPACRDY WTYQGSFTTP PCEECIVWLL LKEPMTVSSD QMAKLRSLLS SAENEPPVPL VSNWRPPQPI NNRVVRASFK
Human
ELISA, SDS-Page
761
Carbonic Anhydrase III/CA3 Protein
>90%
−80°C. Avoid freeze-thaw cycles.