Learn More
Novus Biologicals™ Human CCL11/Eotaxin Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130311
Catalog No. NBP130311
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 24-97 of Human CCL11/Eotaxin The Recombinant Human CCL11/Eotaxin Protein is derived from E. coli. The Recombinant Human CCL11/Eotaxin Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_002977
20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT, 0.1 M NaCl
10.6kDa
Protein
MGSSHHHHHH SSGLVPRGSH MGPASVPTTC CFNLANRKIP LQRLESYRRI TSGKCPQKAV IFKTKLAKDI CADPKKKWVQ DSMKYLDQKS PTPKP
Human
ELISA, SDS-Page
6356
CCL11/Eotaxin protein
>90%
−80°C. Avoid freeze-thaw cycles.