Learn More
Novus Biologicals™ Human CCL17/TARC Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130201
Catalog No. NBP130201
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 24-94 of Human CCL17/TARC The Recombinant Human CCL17/TARC Protein is derived from E. coli. The Recombinant Human CCL17/TARC Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_002978
PBS (pH 7.4), 10% glycerol
10.3kDa
Protein
MGSSHHHHHH SSGLVPRGSH MARGTNVGRE CCLEYFKGAI PLRKLKTWYQ TSEDCSRDAI VFVTVQGRAI CSDPNNKRVK NAVKYLQSLE RS
Human
ELISA, SDS-Page
6361
CCL17/TARC protein
>95%
−80°C. Avoid freeze-thaw cycles.