Learn More
Novus Biologicals™ Human CCL21/6Ckine Protein
Manufacturer:  Novus Biologicals™ NBP137092
Catalog No. NBP137092
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The purity of this protein is >90% by SDS-PAGE. Molecular weight is 14.5kDa (132aa), confirmed by MALDI-TOF.
Specifications
Specifications
NP_002980
This product is lyophilized and contains PBS and 2% Sucrose.
14.5kDa
Protein
MGSSHHHHHH SSGLVPRGSH MSDGGAQDCC LKYSQRKIPA KVVRSYRKQE PSLGCSIPAI LFLPRKRSQA ELCADPKELW VQQLMQHLDK TPSPQKPAQG CRKDRGASKT GKKGKGSKGC KRTERSQTPK GP
Human
ELISA, SDS-Page
6366
CCL21/6Ckine protein
>90%
−80°C. Avoid freeze-thaw cycles.