Learn More
Novus Biologicals™ Human CCL22/MDC Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130245
Catalog No. NBP130245
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 25-93 of Human CCL22/MDC The Recombinant Human CCL22/MDC Protein is derived from E. coli. The Recombinant Human CCL22/MDC Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_002981
PBS (pH 7.4), 10% glycerol
10.3kDa
Protein
MGSSHHHHHH SSGLVPRGSH MGPYGANMED SVCCRDYVRY RLPLRVVKHF YWTSDSCPRP GVVLLTFRDK EICADPRVPW VKMILNKLSQ
Human
ELISA, SDS-Page
6367
CCL22/MDC Protein
>95%
−80°C. Avoid freeze-thaw cycles.