Learn More
Novus Biologicals™ Human CCL26/Eotaxin-3 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130217
Catalog No. NBP130217
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 24-94 of Human CCL26/Eotaxin-3 The Recombinant Human CCL26/Eotaxin-3 Protein is derived from E. coli. The Recombinant Human CCL26/Eotaxin-3 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_006063
PBS (pH 7.4), 10% glycerol
8.5kDa
Protein
MTRGSDISKT CCFQYSHKPL PWTWVRSYEF TSNSCSQRAV IFTTKRGKKV CTHPRKKWVQ KYISLLKTPK QL
Human
ELISA, SDS-Page
10344
CCL26/Eotaxin-3 Protein
>90%
−80°C. Avoid freeze-thaw cycles.