Learn More
Novus Biologicals™ Human CCL27/CTACK Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP141150
Catalog No. NBP141150
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 25-112 of Human CCL27/CTACK The Recombinant Human CCL27/CTACK Protein is derived from E. coli. The Recombinant Human CCL27/CTACK Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_006655
10mM Sodium Citrate buffer (pH 3.5), 10% glycerol
10.3kDa
Protein
MFLLPPSTAC CTQLYRKPLS DKLLRKVIQV ELQEADGDCH LQAFVLHLAQ RSICIHPQNP SLSQWFEHQE RKLHGTLPKL NFGMLRKMG
Human
ELISA, SDS-Page
10850
CCL27/CTACK protein
>95%
−80°C. Avoid freeze-thaw cycles.