Learn More
Novus Biologicals™ Human CCL28 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130232
Catalog No. NBP130232
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 23-127 of Human CCL28 The Recombinant Human CCL28 Protein is derived from E. coli. The Recombinant Human CCL28 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_683513
10mM Sodium Citrate buffer (pH 3.5), 10% glycerol
14.3kDa
Protein
MGSSHHHHHH SSGLVPRGSH MILPIASSCC TEVSHHISRR LLERVNMCRI QRADGDCDLA AVILHVKRRR ICVSPHNHTV KQWMKVQAAK KNGKGNVCHR KKHHGKRNSN RAHQGKHETY GHKTPY
Human
ELISA, SDS-Page
56477
CCL28 Protein
>90%
−80°C. Avoid freeze-thaw cycles.