Learn More
Novus Biologicals™ Human CCL5/RANTES Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130213
Catalog No. NBP130213
Add to cart
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 24-91 of Human CCL5/RANTES The Recombinant Human CCL5/RANTES Protein is derived from E. coli. The Recombinant Human CCL5/RANTES Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_002976
10mM Sodium Citrate buffer (pH 3.5), 10% glycerol
7.9kDa
Protein
MSPYSSDTTP CCFAYIARPL PRAHIKEYFY TSGKCSNPAV VFVTRKNRQV CANPEKKWVR EYINSLEMS
Human
ELISA, SDS-Page
6352
CCL5/RANTES protein
>95%
−80°C. Avoid freeze-thaw cycles.