Learn More
Novus Biologicals™ Human CTGF/CCN2 Protein
Highly purified. Generating reliable and reproducible results. Applications: Functional, SDS-Page
Manufacturer:  Novus Biologicals™ NBP191182
Catalog No. NBP191182
Add to cart
Description
Description
A full-length recombinant protein corresponding to CTGF. Source: E. coli ( NCBI Accession #: NP_001892.) The Recombinant Human CTGF/CCN2 Protein is derived from E. coli. The Recombinant Human CTGF/CCN2 Protein has been validated for the following applications: Functional, SDS-Page.
Specifications
Specifications
P29279
Lyophilized
CTGF/CCN2 protein
−20°C. Avoid freeze-thaw cycles.
Functional, SDS-Page
1490
Protein
Human