Learn More
Novus Biologicals™ Human CX3CL1/Fractalkine Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130220
Catalog No. NBP130220
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 25- 100 of Human CX3CL1/Fractalkine The Recombinant Human CX3CL1/Fractalkine Protein is derived from E. coli. The Recombinant Human CX3CL1/Fractalkine Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_002987
PBS (pH 7.4), 10% glycerol
10.9kDa
Protein
MGSSHHHHHH SSGLVPRGSH MQHHGVTKCN ITCSKMTSKI PVALLIHYQQ NQASCGKRAI ILETRQHRLF CADPKEQWVK DAMQHLDRQA AALTRNG
Human
ELISA, SDS-Page
6376
CX3CL1/Fractalkine protein
>90%
−80°C. Avoid freeze-thaw cycles.