Learn More
Novus Biologicals™ Human Cysteine Dioxygenase Type 1 Recombinant Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130241
Catalog No. NBP130241
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-170 of Human Cysteine Dioxygenase Type 1 The Recombinant Human Cysteine Dioxygenase Type 1 Protein is derived from E. coli. The Recombinant Human Cysteine Dioxygenase Type 1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
AAH_24241
20mM Tris buffer (pH 8.0), 10% glycerol, 1mM DTT
23.9kDa
Protein
0.1mg
Cysteine dioxygenase, 1-170 aa, Human, His-tagged, Recombinant, E.coli (AAH_24241).
RUO
Human
ELISA, SDS-Page
1036
Cysteine Dioxygenase Type 1 Protein
>90%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes