Learn More
Novus Biologicals™ Human Cytosolic Sulfotransferase 2B1/SULT2B1 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP141135
Catalog No. NBP141135
Add to cart
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 1-365 of Human Cytosolic Sulfotransferase 2B1/SULT2B1 The Recombinant Human Cytosolic Sulfotransferase 2B1/SULT2B1 Protein is derived from E. coli. The Recombinant Human Cytosolic Sulfotransferase 2B1/SULT2B1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_814444
20mM Tris-HCl buffer (pH7.5), 10% glycerol
41.3kDa
Protein
MDGPAEPQIP GLWDTYEDDI SEISQKLPGE YFRYKGVPFP VGLYSLESIS LAENTQDVRD DDIFIITYPK SGTTWMIEII CLILKEGDPS WIRSVPIWER APWCETIVGA FSLPDQYSPR LMSSHLPIQI FTKAFFSSKA KVIYMGRNPR DVVVSLYHYS KIAGQLKDPG TPDQFLRDFL KGEVQFGSWF DHIKGWLRMK GKDNFLFITY EELQQDLQGS VERICGFLGR PLGKEALGSV VAHSTFSAMK ANTMSNYTLL PPSLLDHRRG AFLRKGVCGD WKNHFTVAQS EAFDRAYRKQ MRGMPTFPWD EDPEEDGSPD PEPSPEPEPK PSLEPNTSLE REPRPNSSPS PSPGQASETP HPRPS
Human
ELISA, SDS-Page
6820
Cytosolic Sulfotransferase 2B1/SULT2B1 Protein
>85%
−80°C. Avoid freeze-thaw cycles.