Learn More
Novus Biologicals™ Human ErbB2/Her2 Recombinant Protein
Manufacturer:  Novus Biologicals™ NBP194702
Catalog No. NBP194702
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
ELISA
2064
SDS-PAGE
0.05mg
Recombinant human Her-2 encodes amino acid 23 - 652 of the full length protein.
RUO
Human
PBS, pH 7.4.
ErbB2/Her2 protein
>95%
HEK293 cells
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Unlabeled
Yes