Learn More
Novus Biologicals™ Human FGF-10 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP17888650UG
Catalog No. NBP17888650
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 38-208 of Human FGF-10 The Recombinant Human FGF-10 Protein is derived from E. coli. The Recombinant Human FGF-10 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
O15520
Liquid. 20mM Tris-HCl buffer (pH8.0) containing 50% glycerol, 0.2M NaCl, 2mM DTT, 2mM EDTA
22kDa
SDS-PAGE
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
SDS-Page
2255
FGF-10 protein
>95%
Human