Learn More
Novus Biologicals™ Human GABARAP Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP137084
Catalog No. NBP137084
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-117 of Human GABARAP The Recombinant Human GABARAP Protein is derived from E. coli. The Recombinant Human GABARAP Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_009209
20mM Tris-HCl buffer (pH8.0), 20% glycerol, 0.2 M NaCl
16kDa
Protein
MGSSHHHHHH SSGLVPRGSH MKFVYKEEHP FEKRRSEGEK IRKKYPDRVP VIVEKAPKAR IGDLDKKKYL VPSDLTVGQF YFLIRKRIHL RAEDALFFFV NNVIPPTSAT MGQLYQEHHE EDFFLYIAYS DESVYGL
Human
ELISA, SDS-Page
11337
GABARAP Protein
>95%
−80°C. Avoid freeze-thaw cycles.