Learn More
Novus Biologicals™ Human Galectin-1 Recombinant Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130230
Catalog No. NBP130230
Add to cart
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 1-135 of Human Galectin-1 The Recombinant Human Galectin-1 Protein is derived from E. coli. The Recombinant Human Galectin-1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_002296
20mM Tris-HCl buffer (pH 7.5), 10% glycerol
14.7kDa
Protein
0.1mg
Human recombinant Galectin-1, 1-135 aa, purified from E.coli (NP_002296). MACGLVASNL NLKPGECLRV RGEVAPDAKS FVLNLGKDSN NLCLHFNPRF NAHGDANTIV CNSKDGGAWG TEQREAVFPF QPGSVAEVCI TFDQANLTVK LPDGYEFKFP NRLNLEAINY MAADGDFKIK CVAFD
RUO
Human
ELISA, SDS-Page
3956
Galectin-1 Protein
>90%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes