Learn More
Novus Biologicals™ Human Galectin-3 Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
Manufacturer:  Novus Biologicals™ NBP130277
Catalog No. NBP130277
Add to cart
Description
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-250 of Human Galectin-3 The Recombinant Human Galectin-3 Protein is derived from E. coli. The Recombinant Human Galectin-3 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications
Specifications
NP_002297
20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT, 0.1 M NaCl
28.3kDa
Protein
MGSSHHHHHH SSGLVPRGSH MADNFSLHDA LSGSGNPNPQ GWPGAWGNQP AGAGGYPGAS YPGAYPGQAP PGAYPGQAPP GAYPGAPGAY PGAPAPGVYP GPPSGPGAYP SSGQPSATGA YPATGPYGAP AGPLIVPYNL PLPGGVVPRM LITILGTVKP NANRIALDFQ RGNDVAFHFN PRFNENNRRV IVCNTKLDNN WGREERQSVF PFESGKPFKI QVLVEPDHFK VAVNDAHLLQ YNHRVKKLNE ISKLGISGDI DLTSASYTMI
Human
ELISA, SDS-Page
3958
Galectin-3 Protein
>95%
−80°C. Avoid freeze-thaw cycles.