Learn More
Novus Biologicals™ Human Glutathione Reductase Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
Manufacturer:  Novus Biologicals™ NBP17881750UG
Catalog No. NBP17881750
Add to cart
Description
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 43-522 of Human Glutathione Reductase The Recombinant Human Glutathione Reductase Protein is derived from E. coli. The Recombinant Human Glutathione Reductase Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications
Specifications
P00390
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 0.1M NaCl
54.3kDa
SDS-PAGE
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
SDS-Page
2936
Glutathione Reductase Protein
>95%
Human