Learn More
Novus Biologicals™ Human HSP27 Recombinant Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130308
Catalog No. NBP130308
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-205 of Human HSP27 The Recombinant Human HSP27 Protein is derived from E. coli. The Recombinant Human HSP27 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
P04792
20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT, 0.1 M NaCl
24.9kDa
Protein
0.1mg
HSP27, 1-205aa, Human, His tag, E.coli (NP_001531).
RUO
Human
ELISA, SDS-Page
3315
HSP27 Protein
>95%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes